Organic reference standard, Sodium calcium edetate, Ph.Eur., RS
Catalog # EDQMS0700000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:62-33-9
- Description:Sodium calcium edetate
- Formula:C₁₀H₁₂CaN₂Na₂O₈
- Storage temperature:Ambient
- Description:Sodium calcium edetate
Specifications
About this item
- In Monograph 0231
- Soluble in water
- Pharmaceutical applications