Order Entry
Singapore
ContactUsLinkComponent
Organic reference standard, Roxithromycin, Ph.Eur., RS
Organic reference standard, Roxithromycin, Ph.Eur., RS
Catalog # EDQMR1500000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Roxithromycin, Ph.Eur., RS
Catalog # EDQMR1500000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    125 mg
  • Standards:
    Reference standard
  • CAS No.:
    80214-83-1
  • Description:
    Roxithromycin
  • Formula:
    C₄₁H₇₆N₂O₁₅
  • Storage temperature:
    Ambient
  • Description:
    Roxithromycin

Specifications

About this item

Filled under inert atmosphere protected from humidity.

  • In Monograph 1146