Organic reference standard, Retinol ester, Ph.Eur., RS
Catalog # EDQMR0290000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- Description:Retinol ester
- Storage temperature:Ambient
- Description:Retinol ester
Specifications
About this item
- In Monographs 0217, 0218, 0219, and 0220