Organic reference standard, Propyphenazone, Ph.Eur., RS
Catalog # EDQMP3750000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:200 mg
- Standards:Reference standard
- CAS No.:479-92-5
- Description:Propyphenazone
- Formula:C₁₄H₁₈N₂O
- Storage temperature:Ambient
- Description:Propyphenazone
Specifications
About this item
- In Monograph 0636