Order Entry
Singapore
ContactUsLinkComponent
Organic reference standards, Propranolol hydrochloride, Ph.Eur., RS
Organic reference standards, Propranolol hydrochloride, Ph.Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, Propranolol hydrochloride, Ph.Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    318-98-9
  • Formula:
    C₁₆H₂₂ClNO₂
  • Storage temperature:
    Ambient
  • Description:
    Propranolol hydrochloride

Specifications