Order Entry
Singapore
ContactUsLinkComponent
 
Organic reference standard, Promazine hydrochloride, Ph.Eur., RS
Catalog # EDQMP3390000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Promazine hydrochloride, Ph.Eur., RS
Catalog # EDQMP3390000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    53-60-1
  • Description:
    Promazine hydrochloride
  • Formula:
    C₁₇H₂₁ClN₂S
  • Storage temperature:
    Ambient
  • Description:
    Promazine hydrochloride

Specifications

About this item

  • In Monographs 0475 and 1365