Organic reference standard, Primaquine diphosphate, Ph.Eur., RS
Catalog # EDQMP2940000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:500 mg
- Standards:Reference standard
- CAS No.:63-45-6
- Description:Primaquine diphosphate
- Formula:C₁₅H₂₇N₃O₉P₂
- Storage temperature:Ambient
- Description:Primaquine diphosphate
Specifications
About this item
- In Monograph 0635
- Solubility in water: 33,3 to 100 g/l