Organic reference standard, Prilocaine hydrochloride, Ph.Eur., RS
Catalog # EDQMP2939060
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:1786-81-8
- Description:Prilocaine hydrochloride
- Formula:C₁₃H₂₁ClN₂O
- Storage temperature:Ambient
- Description:Prilocaine hydrochloride
Specifications
About this item
- In Monograph 1363
- Soluble in/miscible with DMSO and methanol