Organic reference standard, Phenoxyethanol, Ph.Eur., RS
EDQMP0950000
: EDQM
:
- Pack type:Vial
- Pk:1 mL
- Standards:Reference standard
- CAS No.:122-99-6
- Description:Phenoxyethanol
- Formula:C₈H₁₀O₂
- Storage temperature:Ambient
- Description:Phenoxyethanol
- In Monograph 0781