Organic reference standard, Pentoxifylline, Ph.Eur., RS
EDQMP0510000
: EDQM
:
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:6493-05-6
- Description:Pentoxifylline
- Formula:C₁₃H₁₈N₄O₃
- Storage temperature:Ambient
- Description:Pentoxifylline
- In Monograph 0851