Organic reference standard, Penicillamine disulfide, Ph.Eur., RS
Catalog # EDQMP0320000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:20902-45-8
- Description:Penicillamine disulfide
- Formula:C₁₀H₂₀N₂O₄S₂
- Storage temperature:Ambient
- Description:Penicillamine disulfide
Specifications
About this item
- In Monograph 0566
- Solubility in water: 100 to 1000 g/l