Organic reference standards, Ondansetron, Ph.Eur., RS
Catalog # EDQMY0000196
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:30 mg
- Standards:Reference standard
- CAS No.:103639-04-9
- Description:Ondansetron for TLC system suitability
- Formula:C₁₈H₂₄ClN₃O₃
- Storage temperature:Ambient
- Description:Ondansetron for TLC system suitability
Specifications
About this item
- In Monograph 2016
- Solubility in water: 10 to 33,3 g/l