Organic reference standard, Octyldodecanol, Ph.Eur., RS
EDQMO0101000
: EDQM
:
- Pack type:Vial
- Pk:400 mg
- Standards:Reference standard
- CAS No.:5333-42-6
- Description:Octyldodecanol
- Formula:C₂₀H₄₂O
- Storage temperature:Below 30°C
- Description:Octyldodecanol
- In Monograph 1136