Organic reference standards, Nomegestrol acetate, Ph.Eur., RS
Supplier: EDQM
Certificates
About this item
- In Monograph 1551
- Solubility in water: less than 0,1 g/l
Specifications
- CAS No.:58652-20-3
- Formula:C₂₃H₃₀O₄
- Storage temperature:Ambient
- Description:Nomegestrol acetate