$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standards, Nitrendipine, Ph.Eur., RS
Organic reference standards, Nitrendipine, Ph.Eur., RS
:
EDQM
Organic reference standards, Nitrendipine, Ph.Eur., RS
:
EDQM
In Monograph 1246
CAS No.:
89267-41-4
Formula:
C₁₈H₁₈N₂O₆
Storage temperature:
Ambient
Description:
Nitrendipine impurity A