Organic reference standard, Nimodipine, Ph.Eur., RS
EDQMN0850000
: EDQM
:
- Pack type:Vial
- Pk:50 mg
- Standards:Reference standard
- CAS No.:66085-59-4
- Description:Nimodipine
- Formula:C₂₁H₂₆N₂O₇
- Storage temperature:Ambient
- Description:Nimodipine
- In Monograph 1245
- Soluble in/miscible with chloroform, methanol, and ethylacetate