Order Entry
Singapore
ContactUsLinkComponent
 
Organic reference standard, Naphazoline nitrate, Ph.Eur., RS
Catalog # EDQMN0100000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Naphazoline nitrate, Ph.Eur., RS
Catalog # EDQMN0100000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    10 mg
  • Standards:
    Reference standard
  • CAS No.:
    5144-52-5
  • Description:
    Naphazoline nitrate
  • Formula:
    C₁₄H₁₅N₃O₃
  • Storage temperature:
    Ambient
  • Description:
    Naphazoline nitrate

Specifications

About this item

  • In Monograph 0147
  • Solubility in/miscible with methanol and ethanol