Organic reference standard, Mupirocin lithium, Ph.Eur., RS
Catalog # EDQMM3806000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:73346-79-9
- Description:Mupirocin lithium
- Formula:C₂₆H₄₃LiO₉
- Storage temperature:15 to 25°C
- Description:Mupirocin lithium
Specifications
About this item
- In Monographs 1450 and 1451