Organic reference standards, Metronidazole, Ph.Eur., RS
Supplier: EDQM
Certificates
About this item
- In Monographs 0675 and 0934
- Solubility in water: 1 - 10 g/l
Specifications
- CAS No.:696-23-1
- Formula:C₄H₅N₃O₂
- Storage temperature:Ambient
- Description:Metronidazole impurity A