Organic reference standard, Metoprolol impurity A, Ph.Eur., RS
Catalog # EDQMY0000145
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:10 mg
- Standards:Reference standard
- CAS No.:109632-08-8
- Description:Metoprolol impurity A
- Formula:C₁₄H₂₃NO₃
- Storage temperature:Ambient
- Description:Metoprolol impurity A
Specifications
About this item
- In Monographs 1028 and 1448