Organic reference standard, Methyltestosterone, Ph.Eur., RS
EDQMM1800000
: EDQM
:
- Pack type:Vial
- Pk:10 mg
- Standards:Reference standard
- CAS No.:58-18-4
- Description:Methyltestosterone
- Formula:C₂₀H₃₀O₂
- Storage temperature:Ambient
- Description:Methyltestosterone
Filled under inert atmosphere protected from humidity.
- In Monograph 0410
- Solubility in water: less than 0,1 g/l