Organic reference standard, Methylprednisolone, Ph.Eur., RS
Catalog # EDQMM1750000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:83-43-2
- Description:Methylprednisolone
- Formula:C₂₂H₃₀O₅
- Storage temperature:-20°C ± 5°C
- Description:Methylprednisolone
Specifications
About this item
- In Monographs 0312 and 0561
- Solubility in water: less than 1 g/l
- Shipped in ice