Organic reference standard, Methyl ricinoleate, Ph.Eur., RS
Catalog # EDQMM1770180
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:125 mg
- Standards:Reference standard
- CAS No.:141-24-2
- Description:Methyl ricinoleate
- Formula:C₁₉H₃₆O₃
- Storage temperature:Ambient
- Description:Methyl ricinoleate
Specifications
About this item
- In Monographs 0051 and 2367
- Soluble in/miscible with alcohol and ether