Order Entry
Singapore
ContactUsLinkComponent
 
Organic reference standard, Methotrexate impurity D, Ph.Eur., RS
Catalog # EDQMM1000008
Supplier:  EDQM
CAS Number:  
Organic reference standard, Methotrexate impurity D, Ph.Eur., RS
Catalog # EDQMM1000008
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    10 mg
  • Standards:
    Reference standard
  • CAS No.:
    5623-18-7
  • Description:
    Methotrexate impurity D
  • Formula:
    C₁₅H₁₂N₆O₃
  • Storage temperature:
    Ambient
  • Description:
    Methotrexate impurity D

Specifications

About this item

Filled under inert atmosphere protected from humidity.

  • In Monograph 0560
  • Solubility in water: less than 0,1 g/l