Order Entry
Singapore
ContactUsLinkComponent
 
Organic reference standard, Mesna impurity C, Ph.Eur., RS
Catalog # EDQMY0000316
Supplier:  EDQM
CAS Number:  
Organic reference standard, Mesna impurity C, Ph.Eur., RS
Catalog # EDQMY0000316
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    10 mg
  • Standards:
    Reference standard
  • CAS No.:
    69536-71-6
  • Description:
    Mesna impurity C
  • Formula:
     C₄H₈O₄S₂
  • Storage temperature:
    Ambient
  • Description:
    Mesna impurity C

Specifications

About this item

  • In Monograph 1674
  • Pharmaceutical applications