Organic reference standard, Mefloquine hydrochloride, Ph.Eur., RS
Catalog # EDQMM0253000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:51773-92-3
- Description:Mefloquine hydrochloride
- Formula:C₁₇H₁₇ClF₆N₂O
- Storage temperature:Ambient
- Description:Mefloquine hydrochloride
Specifications
About this item
- In Monograph 1241