Organic reference standard, Mefenamic acid, Ph.Eur., RS
Catalog # EDQMM0252600
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:50 mg
- Standards:Reference standard
- CAS No.:61-68-7
- Description:Mefenamic acid
- Formula:C₁₅H₁₅NO₂
- Storage temperature:Ambient
- Description:Mefenamic acid
Specifications
About this item
- In Monograph 1240