Organic reference standards, Liothyronine, Ph.Eur., RS
EDQML0700000
: EDQM
:
- Pack type:Vial
- Pk:60 mg
- Standards:Reference standard
- CAS No.:55-06-1
- Description:Liothyronine sodium
- Formula:C₁₅H₁₁I₃NNaO₄
- Storage temperature:-20°C ± 5°C
- Description:Liothyronine sodium
- In Monographs 0401 and 0728