Organic reference standard, Levothyroxine sodium, Ph.Eur., RS
Catalog # EDQML0570000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:25416-65-3
- Description:Levothyroxine sodium
- Formula:C₁₅H₁₂I₄NNaO₅
- Storage temperature:-20°C ± 5°C
- Description:Levothyroxine sodium
Specifications
About this item
Filled under inert atmosphere and controlled humidity.
- In Monographs 0401 and 0728
- Solubility in water: 0,1 - 1 g/l