Organic reference standard, Levomepromazine maleate, Ph.Eur., RS
Catalog # EDQML0505000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:50 mg
- Standards:Reference standard
- CAS No.:7104-38-3
- Description:Levomepromazine maleate
- Formula:C₂₃H₂₈N₂O₅S
- Storage temperature:Ambient
- Description:Levomepromazine maleate
Specifications
About this item
- In Monograph 0925