Organic reference standard, Levomepromazine hydrochloride, Ph.Eur., RS
Catalog # EDQML0500000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:1236-99-3
- Description:Levomepromazine hydrochloride
- Formula:C₁₉H₂₅ClN₂OS
- Storage temperature:Ambient
- Description:Levomepromazine hydrochloride
Specifications
About this item
- In Monograph 0505