Organic reference standard, Isosorbide-2-nitrate, Ph.Eur., RS
Catalog # EDQMI0775020
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:50 mg
- Standards:Reference standard
- CAS No.:16106-20-0
- Description:Isosorbide-2-nitrate
- Formula:C₆H₉NO₆
- Storage temperature:Ambient
- Description:Isosorbide-2-nitrate
Specifications
About this item
- In Monographs 1117 and 1118
- Solubility in water: more than 100 g/l