Organic reference standard, Isomalt, Ph.Eur., RS
Catalog # EDQMI0465000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:600 mg
- Standards:Reference standard
- CAS No.:64519-82-0
- Description:Isomalt
- Formula:C₁₂H₂₄O₁₁
- Storage temperature:Ambient
- Description:Isomalt
Specifications
About this item
- In Monograph 1531
- Pharmaceutical applications