Organic reference standard, Isoconazole nitrate, Ph.Eur., RS
Catalog # EDQMI0380000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:200 mg
- Standards:Reference standard
- CAS No.:24168-96-5
- Description:Isoconazole nitrate
- Formula:C₁₈H₁₅Cl₄N₃O₄
- Storage temperature:Ambient
- Description:Isoconazole nitrate
Specifications
About this item
- In Monograph 1017
- Solubility in water: less than 0,1 g/l