Organic reference standard, Isoconazole, Ph.Eur., RS
Catalog # EDQMI0375000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:200 mg
- Standards:Reference standard
- CAS No.:27523-40-6
- Description:Isoconazole
- Formula:C₁₈H₁₄Cl₄N₂O
- Storage temperature:Ambient
- Description:Isoconazole
Specifications
About this item
- In Monograph 1018