Organic reference standard, Imipramine hydrochloride, Ph.Eur., RS
EDQMI0100000
: EDQM
:
- Pack type:Vial
- Pk:50 mg
- Standards:Reference standard
- CAS No.:113-52-0
- Description:Imipramine hydrochloride
- Formula:C₁₉H₂₅ClN₂
- Storage temperature:Ambient
- Description:Imipramine hydrochloride
- In Monographs 0029, 0408, 0889 and 1787