Organic reference standard, Imipenem, Ph.Eur., RS
Catalog # EDQMI0090000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:74431-23-5
- Description:Imipenem
- Formula:C₁₂H₁₉N₃O₅S
- Storage temperature:Ambient
- Description:Imipenem
Specifications
About this item
- In Monograph 1226
- Solubility in water: more than 5 g/l at 25°C
- Pharmaceutical applications