Organic reference standard, Hexamidine diisetionate, Ph.Eur., RS
Catalog # EDQMH0330000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:20 mg
- Standards:Reference standard
- CAS No.:659-40-5
- Description:Hexamidine diisetionate
- Formula:C₂₄H₃₈N₄O₁₀S₂
- Storage temperature:Ambient
- Description:Hexamidine diisetionate
Specifications
About this item
Filled under inert atmosphere protected from humidity.
- In Monograph 1436