Organic reference standard, Heparin Low-Molecular-Mass for assay BRP, Ph.Eur., RS
Catalog # EDQMH0185000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:1 KIT
- Standards:Reference standard
- Description:Heparin Low-Molecular-Mass for assay BRP
- Storage temperature:Ambient
- Description:Heparin Low-Molecular-Mass for assay BRP
Specifications
About this item
- In Monograph 0828
- Soluble in water
- Pharmaceutical applications