Order Entry
Singapore
ContactUsLinkComponent
Organic reference standard, Glibenclamide, Ph.Eur., RS
Organic reference standard, Glibenclamide, Ph.Eur., RS
Catalog # EDQMG0325000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Glibenclamide, Ph.Eur., RS
Catalog # EDQMG0325000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    10238-21-8
  • Description:
    Glibenclamide
  • Formula:
    C₂₃H₂₈ClN₃O₅S
  • Storage temperature:
    Ambient
  • Description:
    Glibenclamide

Specifications

About this item

  • In Monograph 0718
  • Pharmaceutical applications