Organic reference standard, Formoterol for impurity I identification, Ph.Eur., RS
EDQMY0000339
: EDQM
:
- Pack type:Vial
- Pk:10 mg
- Standards:Reference standard
- CAS No.:43229-80-7
- Description:Formoterol for impurity I identification
- Formula:C₄₂H₅₂N₄O₁₂
- Storage temperature:Ambient
- Description:Formoterol for impurity I identification
- In Monograph 1724
- Solubility in water: 1 - 10 g/l