Organic reference standard, Flurbiprofen impurity A, Ph.Eur., RS
Catalog # EDQMF0285202
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:20 mg
- Standards:Reference standard
- CAS No.:6341-72-6
- Description:Flurbiprofen impurity A
- Formula:C₁₅H₁₄O₂
- Storage temperature:Ambient
- Description:Flurbiprofen impurity A
Specifications
About this item
- In Monograph 1519