Organic reference standard, Fluocortolone pivalate, Ph.Eur., RS
Catalog # EDQMF0230000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:29205-06-9
- Description:Fluocortolone pivalate
- Formula:C₂₇H₃₇FO₅
- Storage temperature:Ambient
- Description:Fluocortolone pivalate
Specifications
About this item
- In Monograph 1212
- Soluble in dichloromethane