Organic reference standards, Flunarizine dihydrochloride, Ph.Eur., RS
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:30484-77-6
- Formula:C₂₆H₂₈Cl₂F₂N₂
- Storage temperature:2 to 8°C
- Description:Flunarizine dihydrochloride for system suitability