Organic reference standard, Flumequine, Ph.Eur., RS
Catalog # EDQMF0189000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:60 mg
- Standards:Reference standard
- CAS No.:42835-25-6
- Description:Flumequine
- Formula:C₁₄H₁₂FNO₃
- Storage temperature:Ambient
- Description:Flumequine
Specifications
About this item
- In Monograph 1517
- Pharmaceutical applications