Organic reference standard, Fenoterol hydrobromide, Ph.Eur., RS
EDQMF0050000
: EDQM
:
- Pack type:Vial
- Pk:50 mg
- Standards:Reference standard
- CAS No.:1944-12-3
- Description:Fenoterol hydrobromide
- Formula:C₁₇H₂₂BrNO₄
- Storage temperature:Ambient
- Description:Fenoterol hydrobromide
Filled under inert atmosphere protected from humidity.
- In Monograph 0901