Organic reference standards, Fenofibrate, Ph.Eur., RS
Supplier: EDQM
Certificates
About this item
- In Monograph 1322
- Pharmaceutical applications
Specifications
- CAS No.:217636-48-1
- Formula:C₂₄H₂₇ClO₆
- Storage temperature:Ambient
- Description:Fenofibrate impurity G