Organic reference standard, Erythropoietin BRP, Ph.Eur., RS
EDQME1515000
: EDQM
:
- Pack type:Vial
- Pk:6 mg
- Standards:Reference standard
- CAS No.:11096-26-7
- Description:Erythropoietin BRP
- Storage temperature:Ambient
- Description:Erythropoietin BRP
- In Monograph 1316
- Pharmaceutical industry applications