Organic reference standard, Ergosterol, Ph.Eur., RS
Catalog # EDQME1100000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:20 mg
- Standards:Reference standard
- CAS No.:57-87-4
- Description:Ergosterol
- Formula:C₂₈H₄₄O
- Storage temperature:-20°C ± 5°C
- Description:Ergosterol
Specifications
About this item
Filled under inert atmosphere protected from humidity.
- In Monograph 0082