Organic reference standard, Epirubicin hydrochloride, Ph.Eur., RS
Catalog # EDQME0550000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:56390-09-1
- Description:Epirubicin hydrochloride
- Formula:C₂₇H₃₀ClNO₁₁
- Storage temperature:-20°C with ice
- Description:Epirubicin hydrochloride
Specifications
About this item
Filled under inert atmosphere protected from humidity.
- In Monographs 0662, 0714, and 1590